Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121072
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121072 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121072 Supplier Invitrogen™ Supplier No. PA5121072
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: THP-1, Mouse spleen, Mouse brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: KVERSHGKVN GKGSLWCVDP EYKPNLIQAL KKQPFSSASS QNGSLSPHYL SSVIKQNQVR NLKESDIDAA AAMMLLNTSI EQGILECEKP LPLKTALQKK RSYGNAFHHP SAVRLQESDS LATSIDPKED HNYSASSMAA QRCASRSSVS SLSSVDEVYE FIPKNSHVGS DGSEGFHSEE DTDVDYEDDP LGDSGYASQP CAKISEKGQS GKKMRKQTCQ EIDEELKEAA GSLLHLAGIR TCLGSLISTA KTQNQKQRKK.

This gene encodes a forkhead domain binding protein and may function in the transcriptional regulation of the human T-cell leukemia virus long terminal repeat.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXN2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.43 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene FOXN2
Gene Accession No. P32314
Gene Alias 3230402J05Rik; 6030465J18Rik; Fkh19; forkhead box N1; forkhead box N2; forkhead box protein N2; forkhead homolog 19; Foxn1; FOXN2; HTLF; Human T-cell leukemia virus enhancer factor; RGD1562291
Gene Symbols FOXN2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 172-431 of human FOXN2 (NP_002149.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14236, 3344
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.