Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXN2 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982702
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2702 Supplier Bioss Supplier No. BS13205R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a forkhead domain binding protein and may function in the transcriptional regulation of the human T-cell leukemia virus long terminal repeat.

Specifications

Antigen FOXN2
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FOXN2
Gene Accession No. P32314
Gene Alias 3230402J05Rik; 6030465J18Rik; Fkh19; forkhead box N1; forkhead box N1-like; forkhead box N2; Forkhead box protein N2; forkhead homolog 19; Foxn1; FOXN2; HTLF; Human T-cell leukemia virus enhancer factor; LOC100350890; RGD1562291
Gene Symbols FOXN2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FOXN2.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3344
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.