Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563686
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563686 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563686 Supplier Invitrogen™ Supplier No. PA563686
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGPVMPPSKK PESSGISVSS GLSQCYGGSG FSKALQEDDD LDFSLPDIRL EEGAMEDEEL T Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.

FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXN3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Foxn3
Gene Accession No. O00409
Gene Alias 5430426H20Rik; AA589593; AW556347; C14orf116; checkpoint suppressor 1; CHES; Ches1; Ches1l; forkhead box N3; forkhead box protein N3; Foxn3; HTLFL1; PRO1635
Gene Symbols Foxn3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human FOXN3. Recombinant protein control fragment (Product #RP-103347).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1112
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.