Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXN3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA590188
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA590188 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA590188 Supplier Invitrogen™ Supplier No. PA590188
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: VRPLPITPIG VTAAMRNGIT SCRMRTESEP SCGSPVVSGD PKEDHNYSSA KSSNARSTSP TSDSISSSSS SADDHYEFAT KGSQEGSEGS EGSFRSHESP SDTEEDDRKH SQKEPKDSLG DSGYASQHKK RQHFAKARKV PSDTLPLKKR RTEKPPESDD EEMKEAAGSL LHLAGIRSCL NNITNRTAKG QKEQKETTKN; Positive Samples: Mouse pancreas; Cellular Location: Nucleus.

FOXN3 is a member of the forkhead/winged helixtranscription factor family. Checkpoints are eukaryotic DNA damage-inducible cell cycle arrests at G1 and G2. Check point suppressor 1 suppresses multiple yeast checkpoint mutationsincluding mec1, rad9, rad53 and dun1 by activating aMEC1-independent checkpoint pathway. Alternative splicing isobserved at the locus, resulting in distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXN3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Foxn3
Gene Accession No. Q499D0
Gene Alias 5430426H20Rik; AA589593; AW556347; C14orf116; checkpoint suppressor 1; CHES; Ches1; Ches1l; forkhead box N3; forkhead box protein N3; Foxn3; HTLFL1; PRO1635
Gene Symbols Foxn3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 269-468 of human FOXN3 (NP_0051882).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 71375
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.