Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXN4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106666
Description
Antibody detects endogenous levels of total FOXN4.
Members of the winged-helix/forkhead family of transcription factors, such as FOXN4, are characterized by a 110-amino acid DNA-binding domain that can fold into a variant of the helix-turn-helix motif consisting of 3 alpha helices flanked by 2 large loops or wings. These transcription factors are involved in a variety of biologic processes as key regulators in development and metabolism (Li et al., 2004 [PubMed 15363391]).[supplied by OMIM, Mar 2008].
Specifications
FOXN4 | |
Polyclonal | |
Unconjugated | |
FOXN4 | |
forkhead box N4; forkhead box protein N4; forkhead/winged helix transcription factor FOXN4; FOXN4; sli; slipjig | |
Rabbit | |
Affinity chromatography | |
RUO | |
116810, 121643, 288736 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8K3Q3, Q96NZ1 | |
FOXN4 | |
A synthesized peptide derived from human FOXN4(Accession Q96NZ1), corresponding to amino acid residues D413-S463. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction