Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXN4 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | FOXN4 |
|---|---|
| Dilution | Western Blot 1.0 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
FOXN4 Polyclonal specifically detects FOXN4 in Human samples. It is validated for Western Blot.Specifications
| FOXN4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| FLJ35967, forkhead box N4, forkhead box protein N4, forkhead/winged helix transcription factor FOXN4 | |
| The immunogen is a synthetic peptide directed towards the middle region of human FOXN4 (NP_998761). Peptide sequence QGNLWEEMKDEGFSLDTLGAFADSPLGCDLGASGLTPASGGSDQSFPDLQ | |
| Affinity purified |
| Western Blot 1.0 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS buffer, 2% sucrose | |
| 121643 | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title