Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxO1/FKHR Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP224857MFV500
Description
FoxO1/FKHR Polyclonal antibody specifically detects FoxO1/FKHR in Human samples. It is validated for Western BlotSpecifications
FoxO1/FKHR | |
Polyclonal | |
Western Blot | |
FKH1, FKHRforkhead box protein O1, forkhead box O1, Forkhead box protein O1A, forkhead homolog in rhabdomyosarcoma, Forkhead in rhabdomyosarcoma, FOXO1Aforkhead, Drosophila, homolog of, in rhabdomyosarcoma | |
A portion of amino acids 250-300 of human FOXO1A protein was used as the immunogen. | |
0.1 mL | |
Apoptosis, Cancer, DNA Repair, Growth and Development, Neuronal Cell Markers, Phospho Specific | |
2308 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction