Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXO1 Monoclonal Antibody (CL13278)
GREENER_CHOICE

Catalog No. PIMA546542
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA546542 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA546542 Supplier Invitrogen™ Supplier No. MA546542
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Highest antigen sequence indentity to the following orthologs: Mouse - 100%, Rat - 100%.

FOXO1 (FKHR, ForKHead Box 01) is a member in a subfamily of the forkhead homeotic gene family of transcription factors. Recent experiments have shown that FOXO1 can act as either a coactivator or a corepressor of nuclear receptor activity that is mediated through the LXXLL motif found in the carboxyl terminal region of the FKHR protein. Association of FOXO1 with PAX3 has been implicated in alveolar rhabdomyosarcoma. Recent studies link the anti-tumor activity of FOXO1, the process of autophagy and myogenic growth and differentiation. FOXO1 is the main target of insulin signaling that regulated the metabolic homeostasis in response to oxidative stress. FOXO1 binds to the insulin response element (IRE) with consensus sequence 5'-TT[G/A]TTTTG-3' and the related Daf-16 family binding element (DBE) with consensus sequence 5'- TT[G/A]TTTAC-3'. FOXO1 is a regulator of redox balance, osteoblast numbers and controls bone mass. Further, FOXO1 orchestrates the endocrine function of the skeleton in regulating glucose metabolism, and suppresses the transcriptional activity of RUNX2, an upstream activator of osteocalcin/BGLAP. In hepatocytes, FOXO1 promotes gluconeogenesis by acting together with PPARGC1A to activate the expression of genes such as IGFBP1, G6PC and PPCK1. FOXO1 is an important regulator of cell death acting downstream of CDK1, PKB/AKT1 and SKT4/MST1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXO1
Applications Western Blot
Classification Monoclonal
Clone CL13278
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene FOXO1
Gene Accession No. Q12778
Gene Alias Afxh; AI876417; FKH1; FKHR; Fkhr1; forkhead box O1; forkhead box O1 a; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box protein O1; forkhead box protein O1; LOW QUALITY PROTEIN: forkhead box protein O1; forkhead box protein O1A; Forkhead box protein O1-A; Forkhead in rhabdomyosarcoma; forkhead protein 1; forkhead protein FKHR; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; fox family transcription factor FoxO1a.1; FOX01A; foxhead box protein O1-A; foxo1; FOXO1A; FoxO1a.1; member of FKHR-subclass forkhead/winged helix family; zgc:153388
Gene Symbols FOXO1
Host Species Mouse
Immunogen Synthetic Peptide.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2308
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.