Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO1A Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502279496
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-227-9496 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-227-9496 Supplier Bioss Supplier No. BS9439RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

FOXO1A Polyclonal antibody specifically detects FOXO1A in Bovine, Canine, Chicken, Human, Mouse, Porcine, Rat samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen FOXO1A
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene FOXO1
Gene Accession No. A4L7N3, E1BPQ1, G3V7R4, Q12778, Q9R1E0
Gene Alias Afxh; AI876417; FKH1; FKHR; Fkhr1; forkhead box O1; forkhead box O1 a; forkhead box O1a; forkhead box O1A (rhabdomyosarcoma); forkhead box protein O1; forkhead box protein O1; LOW QUALITY PROTEIN: forkhead box protein O1; forkhead box protein O1A; Forkhead box protein O1-A; Forkhead in rhabdomyosarcoma; forkhead protein 1; forkhead protein FKHR; forkhead, Drosophila, homolog of, in rhabdomyosarcoma; fox family transcription factor FoxO1a.1; FOX01A; foxhead box protein O1-A; foxo1; FOXO1A; FoxO1a.1; member of FKHR-subclass forkhead/winged helix family; zgc:153388
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FOXO1A
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2308, 374231, 397077, 477295, 506618, 56458, 84482
Target Species Bovine, Canine, Chicken, Human, Mouse, Pig, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.