Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO3 Monoclonal Antibody (OTI6H9), TrueMAB™, OriGene
SDP

Catalog No. 501682806
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2806 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2806 Supplier OriGene Supplier No. TA809457
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FOXO3 is a ubiquitously expressed 75 kDa protein member of a subfamily of the forkhead homeotic gene family of transcription factors and shuttles between the cytoplasm and nucleus. FOXO transcription factors are key players of cell fate decisions, metabolism, stress resistance, tumor suppression and are regulated by growth factors, oxidative stress or nutrient deprivation. FOXO3 is involved with mTOR in the regulation of autophagy in skeletal muscle, and activates protein degradation in atrophying muscle cells. FOXO3 has also been implicated in several neurodegenerative disorders including aging, neuromuscular disease, systemic lupus erythmatosus, stroke and diabetic complications.

Specifications

Antigen FOXO3
Applications Western Blot
Classification Monoclonal
Clone OTI6H9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene FOXO3
Gene Accession No. O43524
Gene Alias AF6q21, FKHRL1, FKHRL1P2, FOXO2, FOXO3A
Gene Symbols FOXO3
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 374-673 of human FOXO3 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2309
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.