Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXO3, p Ser253 Antibody (SR2309), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32153150UL
Description
FOXO3 Monoclonal antibody specifically detects FOXO3 in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunohistochemistrySpecifications
FOXO3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunohistochemistry | |
SR2309 | |
Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200 | |
AF6q21, AF6q21 protein, DKFZp781A0677, FKHRL1MGC31925, FKHRL1P2, forkhead box O3, forkhead box protein O3, forkhead homolog (rhabdomyosarcoma) like 1, Forkhead in rhabdomyosarcoma-like 1, forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1, FOXO2, FOXO3Aforkhead box O3A, MGC12739 | |
A synthesized peptide derived from human phospho-FOXO3 [Ser253] (Uniprot #: O43524) | |
50 μg | |
Apoptosis, Cancer, DNA Repair, Phospho Specific, Transcription Factors and Regulators | |
2309 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction