Learn More
FOXO3A Mouse anti-Human, Mouse, Clone: 1F12D11, Proteintech

Mouse Monoclonal Antibody
$518.60
Specifications
Antigen | FOXO3A |
---|---|
Clone | 1F12D11 |
Concentration | 1 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-6855
![]() ![]() |
Proteintech Group Inc
664281IG150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The calculated molecular weight of FOXO3 is 71 kDa, but some experiments using a FOXO3a specific antibody showed the expression of a 90 kDa full-length FOXO3a protein (Full) and a 40 kDa processed FOXO3a product (Short).
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.Specifications
FOXO3A | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
AF6q21, AF6q21 protein, FKHRL1, FKHRL1P2, forkhead box O3, Forkhead box protein O3, FOXO2, FOXO3, FOXO3A | |
Foxo3 | |
IgG2a | |
Protein A | |
Antibody |
1F12D11 | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human, Mouse | |
O43524, Q9WVH4 | |
2309, 56484 | |
FOXO3A Fusion Protein Ag1289 | |
Primary | |
-20°C | |
FOXO3 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.