Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXO3A Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA1805

Catalog No. PA1805


Only null left
Edge
Add to Cart

Description

Description

PA1-805 detects FoxO3. PA1-805 has been successfully used in Western blot procedures. In Western blot analysis of rat heart and lung, this antibody detects a ~ 71 kDa protein representing FoxO3. The PA1-805 immunogen is a synthetic peptide corresponding to residues L N(654) V G N F T G A K Q A S S Q S W V(670) of human FOXO3. This sequence is conserved in human, mouse, and rat.

USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

FOXO3A
Polyclonal
Unconjugated
FOXO3
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; C76856; Fkhr2; FKHRL1; FKHRL1P2; forkhead box O3; forkhead box O3a; forkhead box O3A transcription factor; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO2; FOXO3; FOXO-3; FOXO3A
Rabbit
Antigen affinity chromatography
RUO
2309, 294515, 56484
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 1 mg/mL BSA and 0.05% sodium azide
O43524, Q9WVH4
FOXO3
Synthetic peptide corresponding to residues L N(654) V G N F T G A K Q A S S Q S W V(670) of human FOXO3.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.