Learn More
FOXO3A Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 108491AP150UL
Description
The calculated molecular weight of FOXO3 is 71 kDa, but some experiments using a FOXO3a specific antibody showed the expression of a 90 kDa full-length FOXO3a protein (Full) and a 40 kDa processed FOXO3a product (Short).
FoxO (forkhead box O) proteins are a family of transcription factors characterized by the distinct forkhead domain. The FoxO proteins act in the insulin signaling pathway, but FoxO3 has also been shown to play a role in the regulation of apoptosis, cell cycle progression and cancer. Several splice variants of FoxO3 have been reported.Specifications
| FOXO3A | |
| Polyclonal | |
| Unconjugated | |
| FOXO3 | |
| AF6q21, AF6q21 protein, FKHRL1, FKHRL1P2, forkhead box O3, Forkhead box protein O3, FOXO2, FOXO3, FOXO3A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2309, 294515, 56484 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O43524, Q9WVH4 | |
| Foxo3 | |
| FOXO3A Fusion Protein Ag1289 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.