Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXO3A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501983818
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-3818 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-3818 Supplier Bioss Supplier No. BS1548R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.

Specifications

Antigen FOXO3A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene FOXO3
Gene Accession No. O43524
Gene Alias 1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 antibody; AF6q21 protein; AFX; AFX1; Afxh; BOS_9647; C76856; DAF16 antibody; DKFZp781A0677; FkhR; Fkhr2; FKHR2 antibody; FKHRL1; FKHRL1 antibody; FKHRL1P2; FKHRL1P2 antibody; FKHR-LIKE 1; Forkhead Box O1; forkhead box O3; forkhead box O3A; forkhead box O3A transcription factor; Forkhead Box O4; Forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; Forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO1; FOXO2; Foxo3; FOXO-3; FOXO3A; FOXO4; in rhab; MGC120490; MGC12739; MGC31925; MLLT7
Gene Symbols FOXO3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human FOXO3A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2309
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.