Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXO3A Recombinant Rabbit Monoclonal Antibody (BLR140H)
Rabbit Recombinant Monoclonal Antibody
$173.10 - $436.00
Specifications
Antigen | FOXO3A |
---|---|
Clone | BLR140H |
Concentration | 40 μg/mL |
Content And Storage | 4°C |
Applications | Immunoprecipitation, Western Blot |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIMA544467
|
Invitrogen™
MA544467 |
100 μL |
Each for $436.00
|
|
|||||
PIMA544468
|
Invitrogen™
MA544468 |
20 μL |
Each for $173.10
|
|
|||||
Description
FOXO3A Recombinant Monoclonal Antibody for Western Blot, IP
USP48 is a protein containing domains that associate it with the peptidase family C19, also known as family 2 of ubiquitin carboxyl-terminal hydrolases. Family members function as deubiquitinating enzymes, recognizing and hydrolyzing the peptide bond at the C-terminal glycine of ubiquitin. Enzymes in peptidase family C19 are involved in the processing of poly-ubiquitin precursors as well as that of ubiquitinated proteins.Specifications
FOXO3A | |
40 μg/mL | |
Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
RUO | |
BBS with 0.1% BSA and 0.09% sodium azide; pH 7.8-8.6 | |
O43524, Q9WVH4 | |
2309, 56484 | |
Peptide representing a region between residue 450 and 500 of Human Forkhead box protein O3. | |
Antibody |
BLR140H | |
4°C | |
Recombinant Monoclonal | |
Liquid | |
IgG | |
Human, Mouse | |
FOXO3 | |
1110048B16Rik; 2010203A17Rik; AF6q21; AF6q21 protein; C76856; Fkhr2; FKHRL1; FKHRL1P2; forkhead box O3; forkhead box O3a; forkhead box O3A transcription factor; forkhead box protein O3; forkhead homolog (rhabdomyosarcoma) like 1; forkhead in rhabdomyosarcoma-like 1; forkhead protein FKHR2; forkhead, Drosophila, homolog of, in rhabdomyosarcoma-like 1; FOXO2; FOXO3; FOXO-3; FOXO3A | |
FOXO3 | |
Primary | |
purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title