Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXO4 Antibody (1A3), Novus Biologicals™

Rabbit Monoclonal Antibody
$302.50 - $494.50
Specifications
Antigen | FOXO4 |
---|---|
Clone | 1A3 |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Description
FOXO4 Monoclonal antibody specifically detects FOXO4 in Human samples. It is validated for ELISASpecifications
FOXO4 | |
ELISA | |
Monoclonal | |
Purified | |
RUO | |
Human | |
AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog),translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila),translocated to, 7 | |
A synthesized peptide derived from Human FOXO4 [UniProt P98177] | |
Primary | |
Store at -20 to -70C. Avoid freeze-thaw cycles. |
1A3 | |
ELISA | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Phospho Specific | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
4303 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title