Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXO4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB10059745
Description
FOXO4 Polyclonal specifically detects FOXO4 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
FOXO4 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
AFX, AFX1myeloid/lymphoid or mixed-lineage leukemia (trithorax (Drosophila) homolog);translocated to, 7, Fork head domain transcription factor AFX1, forkhead box O4, forkhead box protein O4, MLLT7MGC120490, myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila);translocated to, 7 | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 1 - 3 ug/mL, Peptide ELISA Detection limit 1:64000 | |
NP_005929.2 | |
FOXO4 | |
Peptide with sequence C-EAASQDRMPQDLD corresponding to internal region according to NP_005929.2. | |
0.1 mg | |
Cell Cycle and Replication | |
4303 | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction