Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXO6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107130
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107130 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107130 Supplier Invitrogen™ Supplier No. PA5107130
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sequence homology: Dog: 86%; Guinea Pig: 86%; Human: 86%; Mouse: 100%; Rat: 100%.

FOXO6 is a transcriptional activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXO6
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene FOXO6
Gene Accession No. Q70KY4
Gene Alias forkhead box O6; forkhead box protein O6; Foxo6
Gene Symbols FOXO6
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal region of Mouse FOXO6.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329934
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.