Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXO6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA535117
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535117 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535117 Supplier Invitrogen™ Supplier No. PA535117
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FOXO6 Polyclonal Antibody for Western Blot, ICC/IF

FOXO6 is a transcriptional activator.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXO6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene FOXO6
Gene Accession No. A8MYZ6, Q70KY4
Gene Alias forkhead box O6; forkhead box protein O6; Foxo6
Gene Symbols FOXO6
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 154-181 amino acids from the central region of human FOXO6.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100132074, 329934
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.