Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXP1 Monoclonal Antibody (D6), Invitrogen™

Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA551093
Description
This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| FOXP1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% Proclin 300; pH 7.4 | |
| Q498D1, Q9H334 | |
| FOXP1 | |
| Recombinant Human FOXP1 protein, Asn461-Pro671 (Accession #Q9H334). | |
| 200 μL | |
| Primary | |
| Human, Pig, Rat | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| D6 | |
| Unconjugated | |
| FOXP1 | |
| 12CC4; 3110052D19Rik; 4932443N09Rik; AI461938; AW494214; FLJ23741; Fork head related protein like B; fork head-related protein like B; forkhead box P1; forkhead box protein P1; Forkhead box protein P1 (FOXP1); Forkhead-related transcription factor 1; Foxp1; Glutamine rich factor 1; glutamine-rich factor 1; hFKH1B; HSPC215; mac-1-regulated forkhead; MFH; MGC12942; MGC88572; MGC99551; QRF1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100525716, 27086, 297480 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction