Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXP1 Monoclonal Antibody (JC12 (JC-12)), eBioscience™
GREENER_CHOICE

Catalog No. 5013930
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-139-30 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-139-30 Supplier Invitrogen™ Supplier No. 14996282
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

Description: This JC12 monoclonal antibody reacts with human Foxp1, a member of the Forkhead family of winged helix transcription factors. Foxp1 is expressed in nearly every human tissue, including lymphoid (e.g. tonsil, spleen, and thymus), epithelium, kidney, blood, and motor neurons. Altered Foxp1 expression has been observed in, among others, cancers of the breast, lung, colon, prostate, and pancreas. Moreover, patients with diffuse large B cell lymphomas express high levels of Foxp1, which can be utilized as a prognostic marker for survival. Finally, Foxp1 has been reported to be involved in early B cell development (i.e. pro-B cell to pre-B cell transition) and motor neuron specification and connectivity. Applications Reported: This JC12 antibody has been reported for use in immunoblotting (WB), and immunohistology staining of paraffin embedded tissue sections. Applications Tested: This JC12 antibody has been tested by western blot analysis on cell lysates prepared from Jurkat cells. This antibody can be used at 1-5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone JC12 (JC-12)
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene FOXP1
Gene Accession No. Q9H334
Gene Alias 12CC4; 3110052D19Rik; 4932443N09Rik; AI461938; AW494214; FLJ23741; Fork head related protein like B; fork head-related protein like B; forkhead box P1; forkhead box protein P1; Forkhead box protein P1 (FOXP1); Forkhead-related transcription factor 1; Foxp1; Glutamine rich factor 1; glutamine-rich factor 1; hFKH1B; HSPC215; mac-1-regulated forkhead; MFH; MGC12942; MGC88572; MGC99551; QRF1
Gene Symbols FOXP1
Host Species Mouse
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27086
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.