Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXP1 Mouse anti-Human, Clone:6E4, Abnova™

Catalog No. 89334599 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-599 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-599 Supplier Abnova Corporation Supplier No. MAB17744
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human FOXP1.

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq]

Specifications

Antigen FOXP1
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6E4
Conjugate Unconjugated
Description forkhead box P1
Dilution ELISA (1:10000)
Flow Cytometry (1:200-1:400)
Immunocytochemistry
Immunohistochemistry (1:200-1:1000)
Western Blot (1:500-1:2000)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene FOXP1
Gene Alias 12CC4, FLJ23741, HSPC215, MGC12942, MGC88572, MGC99551, QRF1, hFKH1B
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 481-677 of human FOXP1 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 27086
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.