Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5118588
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5118588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5118588 Supplier Invitrogen™ Supplier No. PA5118588
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total FOXP1.

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Forkhead box P1 protein contains both DNA-binding- and protein-protein binding-domains. This gene may act as a tumor suppressor as it is lost in several tumor types and maps to a chromosomal region (3p14.1) reported to contain a tumor suppressor gene(s). Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXP1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation Dulbecco's PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FOXP1
Gene Accession No. P58462, Q498D1, Q9H334
Gene Alias 12CC4; 3110052D19Rik; 4932443N09Rik; AI461938; AW494214; FLJ23741; Fork head related protein like B; fork head-related protein like B; forkhead box P1; forkhead box protein P1; Forkhead box protein P1 (FOXP1); Forkhead-related transcription factor 1; Foxp1; Glutamine rich factor 1; glutamine-rich factor 1; hFKH1B; HSPC215; mac-1-regulated forkhead; MFH; MGC12942; MGC88572; MGC99551; QRF1
Gene Symbols FOXP1
Host Species Rabbit
Immunogen A synthesized peptide derived from human FOXP1(Accession Q9H334), corresponding to amino acid residues V630-E677.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 108655, 27086, 297480
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.