Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Abnova™

Catalog No. 89032984 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-032-984 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-032-984 Supplier Abnova Corporation Supplier No. PAB30644
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Rabbit polyclonal antibody raised against partial recombinant human FOXP1.

  • High specificity
  • High reactivity

Specifications

Antigen FOXP1
Applications Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against partial recombinant human FOXP1.
Dilution Immunofluorescence (1-4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
Gene Accession No. Q9H334
Gene Alias 12CC4/FLJ23741/HSPC215/MGC12942/MGC88572/MGC99551/QRF1/hFKH1B
Gene Symbols FOXP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to human FOXP1.
Purification Method Antigen affinity purification
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27086
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.