Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP2 Antibody (CL5310), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP26141225UL
Description
FoxP2 Monoclonal specifically detects FoxP2 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
FoxP2 | |
Monoclonal | |
Unconjugated | |
PBS (pH 7.2), containing 40% glycerol with 0.02% Sodium Azide | |
FOXP2 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AQQLVFQQQLLQMQQLQQQQHLLSLQRQGLISIPPGQAALPVQSLPQAGLSPAEIQQLWKEVTGVHSMEDNGIKHGGLDLTTNNSSSTTSSNTSKASPPITHHS | |
25 μL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
CL5310 | |
Immunohistochemistry 1:1000 - 1:2500, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein | |
Mouse | |
Protein A purified | |
RUO | |
93986 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction