Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP2 Antibody (RAB-S249), HRP - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320155H
Description
FoxP2 Monoclonal antibody specifically detects FoxP2 in Human samples. It is validated for ELISASpecifications
FoxP2 | |
Monoclonal | |
HRP | |
CAG repeat protein 44, CAGH44TNRC10forkhead box protein P2, DKFZp686H1726, forkhead box P2, forkhead/winged-helix transcription factor, SPCH1, trinucleotide repeat containing 10, Trinucleotide repeat-containing gene 10 protein | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP2 protein under non-denaturing conditions. | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
RAB-S249 | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
93986 | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction