Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FOXP2 Mouse anti-Human, Clone:2G11B8, Abnova™

Catalog No. 89333598 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-598 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-598 Supplier Abnova Corporation Supplier No. MAB17175
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human FOXP2.

This gene encodes an evolutionarily conserved transcription factor expressed in fetal and adult brain. This transcription factor is a member of the forkhead/winged-helix (FOX) family of transcription factors, and contains a FOX DNA-binding domain and a large polyglutamine tract. Members of the FOX family of transcription factors are regulators of embryogenesis. The product of this gene is thought to be required for proper development of speech and language regions of the brain during embryogenesis. Although a point mutation in this gene has been associated with the KE pedigree segregating developmental verbal dyspraxia, no association between mutations in this gene and another speech disorder, autism, has been found. Multiple alternative transcripts encoding different isoforms have been identified. [provided by RefSeq]

Specifications

Antigen FOXP2
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 2G11B8
Conjugate Unconjugated
Description forkhead box P2
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Immunohistochemistry
Immunocytochemistry
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene FOXP2
Gene Alias CAGH44, DKFZp686H1726, SPCH1, TNRC10
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 641-740 of human FOXP2 from E. coli.
Quantity 100 μg
Gene ID (Entrez) 93986
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.