Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517977
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517977 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517977 Supplier Invitrogen™ Supplier No. PA517977
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.

FOXP2 is a member of the forkhead/winged-helix (FOX) family of transcription factors. It is expressed in fetal and adult brain as well as in several other organs such as the lung and gut. The protein product contains a FOX DNA-binding domain and a large polyglutamine tract and is an evolutionarily conserved transcription factor, which may bind directly to approximately 300 to 400 gene promoters in the human genome to regulate the expression of a variety of genes. This gene is required for proper development of speech and language regions of the brain during embryogenesis, and may be involved in a variety of biological pathways and cascades that may ultimately influence language development. Mutations in this gene cause speech-language disorder 1 (SPCH1), also known as autosomal dominant speech and language disorder with orofacial dyspraxia. Multiple alternative transcripts encoding different isoforms have been identified in this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXP2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FOXP2
Gene Accession No. O15409
Gene Alias 2810043D05Rik; AI449000; CAG repeat protein 44; CAG-16; CAGH44; D0Kist7; DKFZp686H1726; forkhead box P2; forkhead box protein P2; forkhead/winged-helix transcription factor; FOXP2; OTTHUMP00000196932; RGD1559697; SPCH1; TNRC10; trinucleotide repeat containing 10; trinucleotide repeat-containing gene 10 protein
Gene Symbols FOXP2
Host Species Goat
Immunogen Synthetic peptide sequence (REIEEEPLSEDLE) corresponding to the C-terminus amino acids of FOXP2 (aa 703-715).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 93986
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.