Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Antibody (03), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP321485
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
FoxP3 | |
Monoclonal | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
3 | |
Immunohistochemistry 1:100-1:500, Immunohistochemistry-Paraffin 1:100-1:500 | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, a synthetic peptide corresponding to the center region of the human FoxP3. | |
100 μg | |
Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators | |
50943 | |
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction