Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Antibody (16J4G6) - Delta 2 (exon 2 deleted) specific, Novus Biologicals™
SDP

Catalog No. p-4971041 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224953SS 0.025 mg
NBP224953 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224953SS Supplier Novus Biologicals Supplier No. NBP224953SS

Mouse Monoclonal Antibody has been used in 1 publication

FoxP3 Monoclonal specifically detects FoxP3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FOXP3 Delta 2 (exon 2 deleted) specific
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 16J4G6
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin
Gene Accession No. Q9BZS1
Gene Alias AIID, DIETER, Forkhead Box P3, Forkhead Box Protein P3, FOXP3, FOXP3 Delta, FOXP3 Delta2 (exon 2 deleted) specific, FOXP3Delta2, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, PIDX, scurfin, XPID
Gene Symbols FOXP3
Host Species Mouse
Immunogen A synthetic peptide corresponding to the exon 2 deletion site was used to immunize mice.
Purification Method Ammonium sulfate precipitation
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Test Specificity This antibody is reactive to Human.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Precipitation
Isotype IgM
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.