Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Antibody (3G3) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 1 publication
$206.50 - $499.50
Specifications
Antigen | FoxP3 |
---|---|
Clone | 3G3 |
Concentration | 1.0 mg/ml |
Dilution | Western Blot 2 - 5 ug/mL, Flow Cytometry 100 ng/ 10^6 cells in 50 uL |
Applications | Western Blot, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB10056582T
![]() |
Novus Biologicals
NB10056582SS |
0.025 mg |
Each for $206.50
|
|
|||||
NB10056582
![]() |
Novus Biologicals
NB10056582 |
0.1 mg |
Each for $499.50
|
|
|||||
Description
FoxP3 Monoclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry.Specifications
FoxP3 | |
1.0 mg/ml | |
Western Blot, Flow Cytometry | |
Unconjugated | |
Mouse | |
Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators | |
PBS with 0.05% Sodium Azide | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, foxp3 flow, foxp3 flow cytometry, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
FOXP3 | |
IgG1 κ | |
Protein G purified | |
47 kDa |
3G3 | |
Western Blot 2 - 5 ug/mL, Flow Cytometry 100 ng/ 10^6 cells in 50 uL | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Q9BZS1 | |
50943 | |
Full-length His-tagged murine FOXP3 was used as the immunogen. The precise epitope is not known, but it has been mapped to the N-terminal portion of the protein (NP_001107849). | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title