Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Antibody, FITC, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB600245F
Description
FoxP3 Polyclonal specifically detects FoxP3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
FoxP3 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
Q9BZS1 | |
FOXP3 | |
A synthetic peptide made to the C-terminal region of human FOXP3 (between residues 400-431). [UniProt# Q9BZS1] | |
Affinity Purified | |
RUO | |
Primary | |
Human. This antibody does not appear to detect the mouse FOXP3 protein by western blot, but does work on mouse tissue for IHC (see data below). | |
Store at 4C in the dark. |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
FITC | |
PBS with 0.05% Sodium Azide | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
Rabbit | |
47.2 kDa | |
0.1 mL | |
Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Transcription Factors and Regulators | |
50943 | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction