Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Antibody (FOXP3/7990R), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320675100UG
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
FoxP3 | |
Monoclonal | |
0.2 mg/ml | |
Immunohistochemistry-Paraffin 1-2 μg/mL | |
AIID, AIIDMGC141961, DIETER, forkhead box P3, Forkhead Box Protein P3, FOXP3delta7, immune dysregulation, polyendocrinopathy, enteropathy, X-linked, Immunodeficiency, Polyendocrinopathy, Enteropathy, X-Linked, IPEX, JM2, MGC141961, MGC141963, PIDX, PIDXMGC141963, Scurfin, XPID, XPIDpolyendocrinopathy, enteropathy, X-linked | |
Recombinant fragment (around aa300-431) of human FoxP3 protein (exact sequence is proprietary) | |
100 μg | |
Adaptive Immunity, Cell Biology, Chromatin Research, Immunology, Regulatory Immunology, Transcription Factors and Regulators | |
50943 | |
Store at 4&drg;C. Do not freeze. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
FOXP3/7990R | |
Unconjugated | |
10mM PBS with 0.05% BSA | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction