Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Foxp3 Monoclonal Antibody (3G3), APC, Proteintech
SDP

Catalog No. 502256652 Shop All Proteintech Group Inc Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-225-6652 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-225-6652 Supplier Proteintech Group Inc Supplier No. APC65089100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Foxp3 Monoclonal antibody specifically detects Foxp3 in Mouse samples. It is validated for Flow Cytometry

Specifications

Antigen Foxp3
Applications Flow Cytometry
Classification Monoclonal
Clone 3G3
Concentration 0.2 mg/mL
Conjugate APC
Formulation PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Gene PTPRC
Gene Accession No. P06800
Gene Alias AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID
Host Species Mouse
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19264
Content And Storage 4° C, store in dark
Form Liquid
Isotype IgG1
Target Molecule Mouse
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.