Learn More
Invitrogen™ FOXP3 Monoclonal Antibody (4C7)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA515653
Description
MA5-15653 targets FOXP3 in indirect ELISA, WB applications and shows reactivity with Human samples. The MA5-15653 immunogen is purified recombinant fragment of human FOXP3 expressed in E. Coli. . MA5-15653 detects FOXP3 which has a predicted molecular weight of approximately 47.2kDa.
FOXP3 (Forkhead box protein 3) is a member of the forkhead/winged-helix family of transcriptional regulators, highly conserved across mammals, and essential for normal immune homeostasis. FOXP3 is 381 amino acids long, stably and constitutively expressed at a high level in CD25 + CD4 positive regulatory T cells, a low level in CD4-positive/CD25-negative cells, and is absent in CD4-negative/CD8-positive T cells. FOXP3 may be a master regulatory gene, and a more specific marker of regulatory T cells. Defects in the gene encoding FOXP3 protein cause the scurfy phenotype in mice. In humans FOXP3 defects play a role in IPEX syndrome (immune dysfunction, polyendocrinopathy, enteropathy, X-linked syndrome), also known as X-linked autoimmunity-allergic dysregulation (XLAAD) syndrome. Transcript variants of FOXP3 encoding different isoforms have been identified. In human breast and colon cancer cells, expression of FOXP3 is regulated by p53 in response to the DNA damage.
Specifications
FOXP3 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
Q9BZS1 | |
Foxp3 | |
Purified recombinant fragment of human FOXP3 expressed in E. Coli. | |
RUO | |
50943 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Western Blot | |
4C7 | |
Unconjugated | |
Foxp3 | |
AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
Mouse | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.