Learn More
Description
Specifications
Specifications
| Antigen | FoxP3 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 236A/E7 |
| Conjugate | Brilliant Blue 700 (BB700) |
| Formulation | Aqueous buffered solution containing ≤0.09% sodium azide. |
| Gene Alias | Forkhead box protein P3; Scurfin; AIID; IPEX; JM2; DIETER; PIDX; XPID |
| Host Species | Mouse |
| Immunogen | Human full-length FoxP3 Recombinant Protein |
| Quantity | 100 Tests |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.