Learn More
FOXP3 Monoclonal Antibody (236A/E7), eFluor™ 570, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
$176.65 - $535.65
Specifications
| Antigen | FOXP3 |
|---|---|
| Clone | 236A/E7 |
| Concentration | 0.2 mg/mL |
| Applications | Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-3620
|
Invitrogen™
41477780 |
25 μg |
$196.00
$176.65 / Each Save $19.35 10% Off |
|
|||||
50-112-3621
![]()
|
Invitrogen™
41477782 |
100 μg |
$564.00
$535.65 / Each Save $28.35 5% Off |
|
|||||
Description
Description: The 236A/E7 antibody reacts with human Foxp3 protein also known as FORKHEAD BOX P3, SCURFIN, and JM2. Foxp3, a 49-55 kDa protein, is a member of the forkhead/winged-helix family of transcriptional regulators, and was identified as the gene defective in 'scurfy' (sf) mice. Constitutive high expression of Foxp3 mRNA has been shown in CD4+CD25+ regulatory T cells (Treg cells), and ectopic expression of Foxp3 in CD4+CD25- cells imparts a Treg phenotype in these cells. Intracellular staining and flow cytometric analysis of freshly isolated human peripheral blood mononuclear cells (PBMCs) with the 236A/E7 antibody using the Foxp3/Transcription Factor Staining Buffer Set (cat. 00-5523) and protocol reveals staining of the CD4+CD25bright population. The epitope from 236A/E7 is different from that of PCH101 (cat. 72-5776). This antibody has also been shown to recognize rhesus macaque, sooty mangabey and cynomolgus macaque. Applications Reported: This 236A/E7 antibody has been reported for use in immunohistochemical staining of frozen tissue sections, immunohistochemical staining of formalin-fixed paraffin embedded tissue sections, and microscopy. Applications Tested: This 236A/E7 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded human tissue using high or low pH antigen retrieval and can be used at less than or equal to 5 μg/mL.
FOXP3 Monoclonal antibody specifically detects FOXP3 in Human, Non-human Primate, Rhesus Monkey samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).Specifications
| FOXP3 | |
| 0.2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
| Foxp3 | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Foxp3 |
| 236A/E7 | |
| Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunocytochemistry | |
| eFluor 570 | |
| Mouse | |
| Human, Monkey, Rhesus Macaque | |
| Q9BZS1 | |
| 50943, 574303 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.