Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3, Mouse Anti-Human, RB613, Clone: 259D/C7, BD Horizon™
Mouse Monoclonal Antibody
$164.00 - $406.00
Specifications
Antigen | FoxP3 |
---|---|
Clone | 259D/C7 |
Applications | Flow Cytometry (Intracellular Staining) |
Classification | Monoclonal |
Conjugate | RB613 |
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human (QC Testing) samples. It is validated for Flow Cytometry (Intracellular Staining)Specifications
FoxP3 | |
Flow Cytometry (Intracellular Staining) | |
RB613 | |
Mouse | |
Human | |
scurfin; IPEX; JM2; AIID; XPID; DIETER; PIDX | |
IgG1 | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. |
259D/C7 | |
Monoclonal | |
Purified | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
FoxP3 | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title