Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FoxP3 Mouse anti-Human, RB780, Clone: 236A/E7, BD Horizon™

Catalog No. BDB569085 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB569085 100 Tests
BDB569086 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB569085 Supplier BD Biosciences Supplier No. 569085
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen FoxP3
Applications Flow Cytometry, Control
Classification Monoclonal
Clone 236A/E7
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Forkhead box protein P3; Scurfin; AIID; IPEX; JM2; DIETER; PIDX; XPID
Host Species Mouse
Immunogen Human full-length FoxP3 Recombinant Protein
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50943
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.