Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3, Mouse Anti-Human, RY610, 259D/C7, BD Horizon™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 571136
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human (QC Testing), Rhesus,Cynomolgus,Baboon (Tested in Development) samples. It is validated for Flow Cytometry (Intracellular Staining)
Specifications
FoxP3 | |
Monoclonal | |
RY610 | |
scurfin; IPEX; JM2; AIID; XPID; DIETER; PIDX | |
FoxP3 | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG1 |
Flow Cytometry (Intracellular Staining) | |
259D/C7 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. | |
RUO | |
Human, Rhesus Monkey, Cynomolgus, Baboon | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction