Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP3 Polyclonal Antibody, PE, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS10211RPE
This item is not returnable.
View return policy
Description
FoxP3 Polyclonal antibody specifically detects FoxP3 in Human, Mouse, Rat samples. It is validated for Flow CytometrySpecifications
| FoxP3 | |
| Polyclonal | |
| PE | |
| Foxp3 | |
| AIID; DIETER; forkhead box P3; forkhead box protein P3; Forkhead box protein P3 41 kDa form; Forkhead box protein P3, C-terminally processed; forkhead/winged helix transcription factor 3; Foxp3; FOXP3delta7; immune dysregulation, polyendocrinopathy, enteropathy, X-linked; immunodeficiency, polyendocrinopathy, enteropathy, X-linked; IPEX; JM2; MGC141961; MGC141963; PIDX; regulatory protein Foxp3; RGD1562112; RP23-54C14.1; scurfin; scurfy; sf; XPID | |
| KLH conjugated synthetic peptide derived from human FoxP3 | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| Q99JB6, Q9BZS1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20371, 317382, 50943 | |
| -20°C or -80°C if preferred | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction