Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FoxP4 Antibody (RAB-S40), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP320153MFV450
Description
FoxP4 Monoclonal antibody specifically detects FoxP4 in Human samples. It is validated for Flow CytometrySpecifications
FoxP4 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
116113 | |
Store at 4C in the dark. | |
IgG κ |
Flow Cytometry | |
RAB-S40 | |
Flow Cytometry | |
FKHLA, FLJ40908, FLJ44184, fork head-related protein like A, Fork head-related protein-like A, forkhead box P4, forkhead box protein P4, hFKHLA, winged-helix repressor FOXP4 | |
This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP4 protein under non-denaturing conditions. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction