Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXP4 Chimeric Recombinant Rabbit Monoclonal Antibody (RAB-S40)
GREENER_CHOICE

Catalog No. PIMA548342
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA548342 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA548342 Supplier Invitrogen™ Supplier No. MA548342
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Specificity: This antibody recognizes FOXP4 (Forkhead box protein P4). It binds to a folded domain, amino acids 302-541. FOXP4 is a transcriptional repressor that represses lung-specific expression.

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Many members of the forkhead box gene family, including members of subfamily P, have roles in mammalian oncogenesis. This gene may play a role in the development of tumors of the kidney and larynx. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXP4 Chimeric
Applications ELISA, Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone RAB-S40
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Foxp4
Gene Accession No. Q8IVH2
Gene Alias 1200010K03Rik; 2310007G05Rik; FKHLA; FLJ40908; FLJ44184; fork head-related protein like A; fork head-related protein-like A; forkhead box P4; forkhead box protein P4; Foxp4; hFKHLA; mFKHLA; OTTHUMP00000043536; winged-helix repressor FOXP4
Gene Symbols Foxp4
Host Species Rabbit
Immunogen This antibody was obtained by recombinant antibody (rAb) phage display recognizing FOXP4 protein under non-denaturing conditions.
Purification Method Protein A
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116113
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.