Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXP4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518287
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518287 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518287 Supplier Invitrogen™ Supplier No. PA518287
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with bovine, human and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 4,000.

This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Many members of the forkhead box gene family, including members of subfamily P, have roles in mammalian oncogenesis. This gene may play a role in the development of tumors of the kidney and larynx. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXP4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene Foxp4
Gene Accession No. Q8IVH2
Gene Alias 1200010K03Rik; 2310007G05Rik; FKHLA; FLJ40908; FLJ44184; fork head-related protein like A; fork head-related protein-like A; forkhead box P4; forkhead box protein P4; Foxp4; hFKHLA; mFKHLA; OTTHUMP00000043536; winged-helix repressor FOXP4
Gene Symbols Foxp4
Host Species Goat
Immunogen Synthetic peptide sequence (VESASETIRSAPSG) corresponding to the N-terminus amino acids of FOXP4 (aa 3-17).
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116113
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.