Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FOXP4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530908
Description
Recommended positive controls: HeLa, HeLa nucleus, mouse brain. Predicted reactivity: Mouse (94%), Rhesus Monkey (99%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene belongs to subfamily P of the forkhead box (FOX) transcription factor family. Forkhead box transcription factors play important roles in the regulation of tissue- and cell type-specific gene transcription during both development and adulthood. Many members of the forkhead box gene family, including members of subfamily P, have roles in mammalian oncogenesis. This gene may play a role in the development of tumors of the kidney and larynx. Alternative splicing of this gene produces multiple transcript variants, some encoding different isoforms.
Specifications
FOXP4 | |
Polyclonal | |
Unconjugated | |
Foxp4 | |
1200010K03Rik; 2310007G05Rik; FKHLA; FLJ40908; FLJ44184; fork head-related protein like A; fork head-related protein-like A; forkhead box P4; forkhead box protein P4; Foxp4; hFKHLA; mFKHLA; OTTHUMP00000043536; winged-helix repressor FOXP4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116113, 74123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
0.73 mg/mL | |
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8IVH2, Q9DBY0 | |
Foxp4 | |
Recombinant protein fragment corresponding to a region within amino acids 1 and 275 of Human FOXP4. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction