Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXQ1 Antibody (OTI4D9), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272338F
Description
FOXQ1 Monoclonal specifically detects FOXQ1 in Human samples. It is validated for Immunohistochemistry.Specifications
FOXQ1 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
FOXQ1 | |
Recombinant protein fragment corresponding to amino acids 11-295 of human FOXQ1 (NP_150285) produced in E.coli. | |
0.1 mL | |
Primary | |
Human | |
IgG1 |
Immunohistochemistry | |
OTI4D9 | |
Immunohistochemistry | |
forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor | |
Mouse | |
Affinity Purified | |
Cancer | |
94234.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction