Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXQ1 Antibody (PCRP-FOXQ1-2D2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314070H
This item is not returnable.
View return policy
Description
FOXQ1 Monoclonal antibody specifically detects FOXQ1 in Human samples. It is validated for Western Blot, ELISASpecifications
| FOXQ1 | |
| Monoclonal | |
| HRP | |
| forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor | |
| Recombinant full-length human FOXQ1 protein (Uniprot: Q9C009) | |
| 0.1 mL | |
| Cancer | |
| 94234 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot, ELISA | |
| PCRP-FOXQ1-2D2 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction