Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FOXQ1 Antibody (PCRP-FOXQ1-2D2), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314070MFV610
Description
FOXQ1 Monoclonal antibody specifically detects FOXQ1 in Human samples. It is validated for Flow CytometrySpecifications
FOXQ1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
PCRP-FOXQ1-2D2 | |
Flow Cytometry | |
forkhead box Q1, Hepatocyte nuclear factor 3 forkhead homolog 1, HFH-1, HFH1forkhead box protein Q1, HNF-3/forkhead-like protein 1, winged helix/forkhead transcription factor | |
Recombinant full-length human FOXQ1 protein (Uniprot: Q9C009) | |
0.1 mL | |
Cancer | |
94234 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction