Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ FOXQ1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA518178
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA518178 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA518178 Supplier Invitrogen™ Supplier No. PA518178
Add to Cart
Add to Cart

Goat Polyclonal Antibody

This antibody is predicted to react with mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.

FOXQ1 is a member of the FOX gene family, which is characterized by a conserved 110-amino acid DNA-binding motif called the forkhead or winged helix domain. FOX genes are involved in embryonic development, cell cycle regulation, tissue-specific gene expression, cell signaling, and tumorigenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen FOXQ1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene FOXQ1
Gene Accession No. Q9C009
Gene Alias forkhead box protein Q1; forkhead box Q1; FOXQ1; Hepatocyte nuclear factor 3 forkhead homolog 1; Hfh1; HFH-1; Hfh1l; HFH-1l; HNF-3/forkhead homolog 1 like; HNF-3/forkhead homolog-1; HNF-3/forkhead-like protein 1; sa; satin; winged helix/forkhead transcription factor
Gene Symbols FOXQ1
Host Species Goat
Immunogen Synthetic peptide sequence (KLEVFVPRAAHGD) corresponding to the N-terminus amino acids of FOXQ1 (aa 2-15).
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 94234
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.